Gene Transcripts

Gene Symbol
GPATCH3
Entrez Gene ID
63906
Gene Name
G-patch domain containing 3
Gene Aliases
GPATC3
Chromosome Location
Chr. 1: 26890488 - 26900471 on Build GRCh38
UniGene ID
Hs.10903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022078.2 NP_071361.2 7 1414
GenBank mRNA AK022517.1 - 7 1409
AK057930.1 - 7 1409
AK303183.1 - 8 1357
AK308100.1 - 7 1277
AL833886.1 - 7 1396
BC007767.2 - 7 1390 4298740
CR936767.1 - 7 1414

View Full Product Details