Gene Transcripts

Gene Symbol
C1orf50
Entrez Gene ID
79078
Gene Name
chromosome 1 open reading frame 50
Gene Aliases
Chromosome Location
Chr. 1: 42767245 - 42775742 on Build GRCh38
UniGene ID
Hs.148845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024097.3 NP_077002.2 4 393
NR_040733.1 3 277
GenBank mRNA BC001508.2 - 4 369
BC001711.1 - 4 369 3543543
BM702424.1 - 3 229
CN256666.1 - 3 177

View Full Product Details