Gene Transcripts

Gene Symbol
LBHD1
Entrez Gene ID
79081
Gene Name
LBH domain containing 1
Gene Aliases
C11orf48
Chromosome Location
Chr. 11: 62662817 - 62671769 on Build GRCh38
UniGene ID
Hs.9061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024099.3 NP_077004.2 4 696
GenBank mRNA AK055736.1 - 2 2287
AK302538.1 - 4 542
BC001434.2 - 4 696 3138904

View Full Product Details