Gene Transcripts

Gene Symbol
GLMP
Entrez Gene ID
112770
Gene Name
glycosylated lysosomal membrane protein
Gene Aliases
C1orf85, NCU-G1
Chromosome Location
Chr. 1: 156292687 - 156295689 on Build GRCh38
UniGene ID
Hs.202522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256604.1 NP_001243533.1 3 589
NM_001256605.1 NP_001243534.1 2 331
NM_001256608.1 NP_001243537.1 3 589
NM_001256609.1 NP_001243538.1 3 818
NM_144580.2 NP_653181.1 3 589
XM_011509117.2 XP_011507419.1 2 248
XM_017000149.1 XP_016855638.1 3 585
GenBank mRNA AK075349.1 - 3 558
AK296157.1 - 3 786
AL528533.3 - 2 303
AY358450.1 - 3 562
BC011575.1 - 3 572
BC018757.1 - 3 558 4850051
BC036340.1 - 3 505
BP302818.1 - 1 56
CX756681.1 - 3 504

View Full Product Details