Gene Transcripts

Gene Symbol
B4GALNT4
Entrez Gene ID
338707
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Gene Aliases
Chromosome Location
Chr. 11: 369773 - 382117 on Build GRCh38
UniGene ID
Hs.148074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178537.4 NP_848632.2 12, 13 1213
XM_017017654.1 XP_016873143.1 12, 13 2525
GenBank mRNA AB089939.1 - 12, 13 1204

View Full Product Details