Gene Transcripts

Gene Symbol
C11orf53
Entrez Gene ID
341032
Gene Name
chromosome 11 open reading frame 53
Gene Aliases
Chromosome Location
Chr. 11: 111245720 - 111286406 on Build GRCh38
UniGene ID
Hs.298685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198498.2 NP_940900.1 4 998
XM_011542803.2 XP_011541105.1 5 1141
XM_011542804.2 XP_011541106.1 4 924
GenBank mRNA BC039669.1 - 4 998

View Full Product Details