Gene Transcripts

Gene Symbol
C2orf50
Entrez Gene ID
130813
Gene Name
chromosome 2 open reading frame 50
Gene Aliases
Chromosome Location
Chr. 2: 11133053 - 11154564 on Build GRCh38
UniGene ID
Hs.406894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006711859.3 XP_006711922.1 4 1682
GenBank mRNA AK057872.1 - 3 2626

View Full Product Details