Gene Transcripts

Gene Symbol
SCARA5
Entrez Gene ID
286133
Gene Name
scavenger receptor class A member 5
Gene Aliases
NET33, Tesr
Chromosome Location
Chr. 8: 27869882 - 27992852 on Build GRCh38
UniGene ID
Hs.591833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173833.5 NP_776194.2 9 2352
GenBank mRNA AK172746.1 - 9 2182
AY358150.1 - 8 1500
BQ944057.1 - 1 387

View Full Product Details