Gene Transcripts

Gene Symbol
LRRC15
Entrez Gene ID
131578
Gene Name
leucine rich repeat containing 15
Gene Aliases
LIB
Chromosome Location
Chr. 3: 194355247 - 194369743 on Build GRCh38
UniGene ID
Hs.288467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135057.2 NP_001128529.2 3 4915
NM_130830.4 NP_570843.2 2 4858
GenBank mRNA AK022342.1 - 1 2165
AK125112.1 - 2 4858

View Full Product Details