Gene Transcripts
- Gene Symbol
- C6orf89
- Entrez Gene ID
- 221477
- Gene Name
- chromosome 6 open reading frame 89
- Gene Aliases
- BRAP
- Chromosome Location
- Chr. 6: 36834886 - 36928964 on Build GRCh38
- UniGene ID
- Hs.433381
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286635.1 |
NP_001273564.1
|
6, 7 |
990 |
|
NM_001286636.1 |
NP_001273565.1
|
7, 8 |
1054 |
|
NM_001286637.1 |
NP_001273566.1
|
6, 7 |
931 |
|
NM_152734.3 |
NP_689947.2
|
5, 6 |
890 |
|
XM_011514377.2 |
XP_011512679.1
|
8, 9 |
1753 |
|
XM_017010432.1 |
XP_016865921.1
|
8, 9 |
1207 |
|
XM_017010433.1 |
XP_016865922.1
|
8, 9 |
1208 |
|
XM_017010434.1 |
XP_016865923.1
|
7, 8 |
1446 |
|
GenBank mRNA |
AK058086.1 |
- |
7, 8 |
1054 |
|
AK172780.1 |
- |
5, 6 |
799 |
|
AK300351.1 |
- |
6, 7 |
868 |
|
AK315984.1 |
- |
6, 7 |
840 |
|
AY358637.1 |
- |
6, 7 |
864 |
|
BC009380.2 |
- |
1, 2 |
104 |
|
BC023605.2 |
- |
5, 6 |
748 |
|
BC095423.1 |
- |
6, 7 |
903 |
30348810 |
BC105946.1 |
- |
5, 6 |
797 |
40032607 |
BC105947.1 |
- |
5, 6 |
798 |
40032604 |
BC107489.1 |
- |
5, 6 |
798 |
40032608 |
BX538108.1 |
- |
5, 6 |
783 |
|
BX647220.1 |
- |
6, 7 |
862 |
|
FR832871.1 |
- |
6, 7 |
934 |
|
FR832872.1 |
- |
5, 6 |
919 |
|
View Full Product Details