Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037954.3 | NP_001033043.1 | 20 | 4549 | |
NM_033425.4 | NP_219493.1 | 16 | 4082 | ||
XM_017018466.1 | XP_016873955.1 | 21 | 4509 | ||
XM_017018467.1 | XP_016873956.1 | 21 | 4377 | ||
XM_017018468.1 | XP_016873957.1 | 16 | 3693 | ||
XM_017018469.1 | XP_016873958.1 | 17 | 3718 | ||
GenBank mRNA | AB051522.1 | - | 16 | 3989 | |
AF070621.1 | - | 1 | 932 | ||
AK055469.1 | - | 7 | 2861 | ||
AK074277.1 | - | 1 | 2020 | ||
BC033034.2 | - | 16 | 4041 | 4820652 | |
DQ642016.1 | - | 20 | 4549 |