Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052853.3 | NP_443085.2 | 1 | 835 | |
XM_006716170.3 | XP_006716233.1 | 1 | 1060 | ||
XM_011516675.2 | XP_011514977.1 | 1 | 1060 | ||
GenBank mRNA | AF131745.1 | - | 1 | 834 | |
AY302592.1 | - | 1 | 835 | ||
BC014107.2 | - | 1 | 832 | 4138734 |