Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318512.1 | NP_001305441.1 | 5 | 1240 | |
NM_001318514.1 | NP_001305443.1 | 6 | 1400 | ||
NM_001318515.1 | NP_001305444.1 | 3 | 963 | ||
NM_001318516.1 | NP_001305445.1 | 5 | 1207 | ||
NM_019009.3 | NP_061882.2 | 6 | 1390 | ||
XM_017017931.1 | XP_016873420.1 | 4 | 1297 | ||
GenBank mRNA | AB209541.1 | - | 7 | 1095 | |
AK022871.1 | - | 6 | 1297 | ||
AK090850.1 | - | 5 | 1201 | ||
AK096693.1 | - | 5 | 1164 | ||
AK127123.1 | - | 6 | 1353 | ||
AL136835.1 | - | 6 | 1337 | ||
BC004420.1 | - | 6 | 1350 | 3356339 | |
BC012057.1 | - | 6 | 1315 | 4647828 | |
BC018272.1 | - | 6 | 1307 | 3452585 | |
BX648136.1 | - | 1 | 1667 |