Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020243.4 | NP_064628.1 | 3, 4 | 386 | |
GenBank mRNA | AB040119.1 | - | 3, 4 | 368 | |
AB041906.1 | - | 3, 4 | 369 | ||
AF163256.1 | - | 3, 4 | 369 | ||
AK024731.1 | - | 3, 4 | 379 | ||
AK223021.1 | - | 3, 4 | 384 | ||
AK312076.1 | - | 3, 4 | 360 | ||
BC009363.2 | - | 3, 4 | 361 | 4302531 | |
CR456475.1 | - | 3, 4 | 361 |