Gene Transcripts
- Gene Symbol
- BBS7
- Entrez Gene ID
- 55212
- Gene Name
- Bardet-Biedl syndrome 7
- Gene Aliases
- BBS2L1
- Chromosome Location
- Chr. 4: 121824329 - 121870520 on Build GRCh38
- UniGene ID
- Hs.591694
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018190.3 |
NP_060660.2
|
2, 3 |
283 |
|
NM_176824.2 |
NP_789794.1
|
2, 3 |
283 |
|
XM_005263106.3 |
XP_005263163.1
|
2, 3 |
306 |
|
XM_011532079.2 |
XP_011530381.1
|
2, 3 |
306 |
|
XM_011532080.2 |
XP_011530382.1
|
2, 3 |
306 |
|
XM_011532081.2 |
XP_011530383.1
|
2, 3 |
306 |
|
XM_017008357.1 |
XP_016863846.1
|
2, 3 |
306 |
|
XM_017008358.1 |
XP_016863847.1
|
2, 3 |
306 |
|
GenBank mRNA |
AF521643.1 |
- |
2, 3 |
247 |
|
AF521644.1 |
- |
2, 3 |
247 |
|
AK001577.1 |
- |
2, 3 |
247 |
|
AK308076.1 |
- |
2, 3 |
274 |
|
AK309803.1 |
- |
|
247 |
|
BC032691.1 |
- |
2, 3 |
200 |
5240398 |
BG720664.1 |
- |
2, 3 |
278 |
|
BQ940332.1 |
- |
2, 3 |
248 |
|
BX510161.1 |
- |
2, 3 |
284 |
|
View Full Product Details