Gene Transcripts

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
Chromosome Location
Chr. 4: 25377226 - 25418498 on Build GRCh38
UniGene ID
Hs.152173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286756.1 NP_001273685.1 8, 9 735
NM_013367.2 NP_037499.2 8, 9 735
XM_011513838.1 XP_011512140.1 8, 9 612
XM_011513839.1 XP_011512141.1 8, 9 656
XM_017008138.1 XP_016863627.1 8, 9 656
GenBank mRNA AF191338.1 - 7, 8 606
AK023597.1 - 5, 6 346
AK292336.1 - 8, 9 655
AY429517.1 - 1, 2 83
AY429518.1 - 1, 2 70
BC059383.1 - 8, 9 708 30349524

View Full Product Details