Gene Transcripts

Gene Symbol
POMP
Entrez Gene ID
51371
Gene Name
proteasome maturation protein
Gene Aliases
C13orf12, HSPC014, PNAS-110, UMP1
Chromosome Location
Chr. 13: 28659004 - 28678957 on Build GRCh38
UniGene ID
Hs.268742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015932.5 NP_057016.1 6 1201
GenBank mRNA BC003390.1 - 6 1064
BC014334.1 - 6 1095 4045091

View Full Product Details