Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318151.1 | NP_001305080.1 | 3 | 476 | |
NM_001318152.1 | NP_001305081.1 | 2 | 446 | ||
NM_003776.3 | NP_003767.2 | 3 | 826 | ||
GenBank mRNA | AF034091.1 | - | 3 | 185 | |
AJ295637.1 | - | 3 | 183 | ||
AK057708.1 | - | 1 | 2212 | ||
AK123768.1 | - | 3 | 826 | ||
BC009707.1 | - | 3 | 215 | 3876531 | |
BP309678.1 | - | 2 | 446 | ||
BX382916.2 | - | 3 | 400 | ||
CR456532.1 | - | 3 | 178 |