Gene Transcripts
- Gene Symbol
- EMILIN1
- Entrez Gene ID
- 11117
- Gene Name
- elastin microfibril interfacer 1
- Gene Aliases
- EMI, EMILIN, gp115
- Chromosome Location
- Chr. 2: 27078567 - 27086403 on Build GRCh38
- UniGene ID
- Hs.63348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007046.3 |
NP_008977.1
|
2, 3 |
791 |
|
GenBank mRNA |
AF088916.1 |
- |
2, 3 |
292 |
|
AK075517.1 |
- |
2, 3 |
716 |
|
AK307539.1 |
- |
2, 3 |
720 |
|
AL050138.1 |
- |
2, 3 |
222 |
|
AL552691.3 |
- |
2, 3 |
797 |
|
BC007530.1 |
- |
2, 3 |
463 |
|
BC136279.1 |
- |
2, 3 |
391 |
9020266 |
BC142638.1 |
- |
2, 3 |
724 |
|
BC142688.1 |
- |
2, 3 |
673 |
40148654 |
View Full Product Details