Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003825.3 | NP_003816.2 | 8 | 1527 | |
NM_130798.2 | NP_570710.1 | 7 | 1368 | ||
XM_006720725.2 | XP_006720788.1 | 9 | 1606 | ||
GenBank mRNA | AJ011915.1 | - | 8 | 1149 | |
BC000148.2 | - | 8 | 1159 | 2900640 | |
BC003686.1 | - | 8 | 1178 | 3461227 | |
BC022890.1 | - | 8 | 1184 | 4658930 |