Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
8800
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
PEX11-ALPHA, PMP28, hsPEX11p
Chromosome Location
Chr. 15: 89681531 - 89690784 on Build GRCh38
UniGene ID
Hs.31034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271572.1 NP_001258501.1 3 982
NM_001271573.1 NP_001258502.1 2 959
NM_003847.2 NP_003838.1 3 1075
GenBank mRNA AB015594.1 - 3 1018
AF093668.1 - 3 980
AK001415.1 - 3 1068
AK297442.1 - 2 807
AK300978.1 - 3 860
AL360141.1 - 1 189
BC009697.1 - 3 969 3878406

View Full Product Details