Gene Transcripts

Gene Symbol
GYG2
Entrez Gene ID
8908
Gene Name
glycogenin 2
Gene Aliases
GN-2, GN2
Chromosome Location
Chr. X: 2828712 - 2882820 on Build GRCh38
UniGene ID
Hs.567381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079855.1 NP_001073324.1 3 308
NM_001184702.1 NP_001171631.1 3 308
NM_001184703.1 NP_001171632.1 4 401
NM_003918.2 NP_003909.2 4 401
XM_011545600.2 XP_011543902.1 3 239
XM_017029927.1 XP_016885416.1 4 351
XM_017029928.1 XP_016885417.1 4 326
XM_017029929.1 XP_016885418.1 4 511
XM_017029930.1 XP_016885419.1 3 216
XM_017029931.1 XP_016885420.1 3 239
GenBank mRNA AK292496.1 - 3 216
BC023152.1 - 3 175 3923041
DQ424901.1 - 2 119
U94362.1 - 4 402
U94363.1 - 3 182

View Full Product Details