Gene Transcripts

Gene Symbol
ZCCHC14
Entrez Gene ID
23174
Gene Name
zinc finger CCHC-type containing 14
Gene Aliases
BDG-29, BDG29
Chromosome Location
Chr. 16: 87406246 - 87492948 on Build GRCh38
UniGene ID
Hs.156231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015144.2 NP_055959.1 12, 13 2829
GenBank mRNA AB011151.2 - 12, 13 2687
AB030243.1 - 12, 13 2837
AK289682.1 - 12, 13 3020
BC101478.1 - 12, 13 2836 8068984

View Full Product Details