Gene Transcripts

Gene Symbol
RPL14
Entrez Gene ID
9045
Gene Name
ribosomal protein L14
Gene Aliases
CAG-ISL-7, CTG-B33, L14, RL14, hRL14
Chromosome Location
Chr. 3: 40457292 - 40462372 on Build GRCh38
UniGene ID
Hs.446522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034996.2 NP_001030168.1 5, 6 497
NM_003973.4 NP_003964.3 5, 6 433
GenBank mRNA AK222750.1 - 1 392
AK223111.1 - 5, 6 386
AK290674.1 - 5, 6 393
AK300873.1 - 5, 7 386
BC000606.2 - 5, 6 399 3346054
BC005134.2 - 4, 5 364 3928410
BC009294.2 - 5, 6 396 4123062
BC019651.1 - 1 371 4454086
BC022805.1 - 1 361
BC029036.1 - 5, 6 485 5183728
BC071913.1 - 1 393 6671085
BG564298.1 - 5, 6 433
BG776586.1 - 5, 6 416
CD298951.1 - 5, 6 500
D87735.1 - 5, 6 381
DQ118667.1 - 4, 5 364
L10376.1 - 1 369
U16738.1 - 4, 5 370
U92982.1 - 4, 5 298

View Full Product Details