Gene Transcripts

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
Chromosome Location
Chr. 16: 68022944 - 68079323 on Build GRCh38
UniGene ID
Hs.534460
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271762.1 NP_001258691.1 9 934
NM_001271763.1 NP_001258692.1 8 909
NM_017803.4 NP_060273.1 10 1014
GenBank mRNA AB101210.1 - 10 678
AK000406.1 - 9 581
AK290578.1 - 10 716
AK296663.1 - 8 561
BC006527.2 - 10 657 2988128

View Full Product Details