Gene Transcripts
- Gene Symbol
- APPL2
- Entrez Gene ID
- 55198
- Gene Name
- adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
- Gene Aliases
- DIP13B
- Chromosome Location
- Chr. 12: 105173297 - 105236230 on Build GRCh38
- UniGene ID
- Hs.506603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001251904.1 |
NP_001238833.1
|
7 |
661 |
|
NM_001251905.1 |
NP_001238834.1
|
6, 7 |
686 |
|
NM_018171.3 |
NP_060641.2
|
6, 7 |
643 |
|
XM_006719472.1 |
XP_006719535.1
|
7 |
580 |
|
XM_011538530.2 |
XP_011536832.1
|
7 |
2113 |
|
XM_011538531.2 |
XP_011536833.1
|
7 |
1776 |
|
XM_011538532.2 |
XP_011536834.1
|
7 |
3102 |
|
XM_017019551.1 |
XP_016875040.1
|
6, 7 |
1048 |
|
XM_017019552.1 |
XP_016875041.1
|
6, 7 |
1758 |
|
XM_017019553.1 |
XP_016875042.1
|
7, 8 |
3356 |
|
XM_017019554.1 |
XP_016875043.1
|
6, 7 |
562 |
|
GenBank mRNA |
AK001521.2 |
- |
6, 7 |
463 |
|
AK290562.1 |
- |
6, 7 |
496 |
|
AK293783.1 |
- |
6, 7 |
435 |
|
AK296610.1 |
- |
6, 7 |
442 |
|
AK297100.1 |
- |
7 |
514 |
|
AK310377.1 |
- |
6, 7 |
431 |
|
AY113704.1 |
- |
6, 7 |
643 |
|
BC033731.1 |
- |
6, 7 |
547 |
5168590 |
View Full Product Details