Gene Transcripts

Gene Symbol
C12orf43
Entrez Gene ID
64897
Gene Name
chromosome 12 open reading frame 43
Gene Aliases
Chromosome Location
Chr. 12: 121002422 - 121016502 on Build GRCh38
UniGene ID
Hs.528671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286191.1 NP_001273120.1 4 447
NM_001286192.1 NP_001273121.1 4 354
NM_001286195.1 NP_001273124.1 4 354
NM_001286196.1 NP_001273125.1 4 354
NM_001286197.1 NP_001273126.1 3 255
NM_001286198.1 NP_001273127.1 5 611
NM_022895.2 NP_075046.1 4 354
NR_104409.1 4 364
XM_017019829.1 XP_016875318.1 4 438
GenBank mRNA AK022510.1 - 4 349
AK225162.1 - 4 349
AK299170.1 - 5 597
AK301566.1 - 4 339
AK307971.1 - 4 449
BC014661.1 - 4 328 4860511
BP201132.1 - 4 334
BP343829.1 - 3 182
DA014862.1 - 4 353
DC297725.1 - 4 341

View Full Product Details