Gene Transcripts

Gene Symbol
IQCK
Entrez Gene ID
124152
Gene Name
IQ motif containing K
Gene Aliases
Chromosome Location
Chr. 16: 19716456 - 19858467 on Build GRCh38
UniGene ID
Hs.460217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305121.1 NP_001292050.1 3 933
NM_001305122.1 NP_001292051.1 4 998
NM_153208.2 NP_694940.1 4 998
NR_130967.1 4 998
NR_130968.1 4 998
XM_011545736.2 XP_011544038.1 3 333
XM_017022933.1 XP_016878422.1 3 1044
GenBank mRNA AF520569.1 - 4 998
AF520570.1 - 4 998
AK000122.1 - 3 933
AK093894.1 - 3 269
AK301933.1 - 3 321
AK303998.1 - 2 226
AK311216.1 - 3 291
AK315672.1 - 3 321
BC034823.1 - 3 329 5165639

View Full Product Details