Gene Transcripts

Gene Symbol
DNM1P46
Entrez Gene ID
196968
Gene Name
dynamin 1 pseudogene 46
Gene Aliases
C15orf51, DNM1DN14.2, DNM1DN14@
Chromosome Location
Chr. 15: 99790156 - 99806927 on Build GRCh38
UniGene ID
Hs.567763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003260.1 5 2663
GenBank mRNA AK127834.1 - 5 2663
AK302717.1 - 5 2464
AL833912.1 - 4 2518

View Full Product Details