Gene Transcripts

Gene Symbol
SEPT5-GP1BB
Entrez Gene ID
100526833
Gene Name
SEPT5-GP1BB readthrough
Gene Aliases
Chromosome Location
Chr. 22: 19717220 - 19724774 on Build GRCh38
UniGene ID
Hs.283743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037611.1 11 3156
NR_037612.1 11 1660
GenBank mRNA AB209168.1 - 11 3156
BC020397.1 - 1 41
BC024304.1 - 4 817
L20860.1 - 1 392
U59632.1 - 11 1659

Gene Symbol
SEPT5
Entrez Gene ID
5413
Gene Name
septin 5
Gene Aliases
CDCREL, CDCREL-1, CDCREL1, H5, HCDCREL-1, PNUTL1
Chromosome Location
Chr. 22: 19714464 - 19723322 on Build GRCh38
UniGene ID
Hs.728762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009939.2 NP_001009939.1 11 1930
NM_002688.5 NP_002679.2 12 1736
GenBank mRNA AK056273.1 - 11 1907
BC025261.1 - 12 1700 4622405
U74628.1 - 12 1698
Y11593.1 - 12 1656

View Full Product Details