Gene Transcripts

Gene Symbol
NAP1L3
Entrez Gene ID
4675
Gene Name
nucleosome assembly protein 1 like 3
Gene Aliases
MB20, NPL3
Chromosome Location
Chr. X: 93670926 - 93673683 on Build GRCh38
UniGene ID
Hs.21365
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004538.5 NP_004529.2 1 602
GenBank mRNA AK090772.1 - 2 408
AK095682.1 - 2 459
AK223621.1 - 1 533
AK315176.1 - 1 495
BC034954.2 - 1 490 4823724
D50370.1 - 1 488
DA486883.1 - 1 118

View Full Product Details