Gene Transcripts
- Gene Symbol
- ASMTL
- Entrez Gene ID
- 8623
- Gene Name
- acetylserotonin O-methyltransferase-like
- Gene Aliases
- ASMTLX, ASMTLY, ASTML
- Chromosome Location
- Chr. X: 1403139 - 1453794 on Build GRCh38
- UniGene ID
- Hs.533514
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001173473.1 |
NP_001166944.1
|
8, 9 |
1120 |
|
NM_001173474.1 |
NP_001166945.1
|
7, 8 |
1157 |
|
NM_004192.3 |
NP_004183.2
|
8, 9 |
1205 |
|
XM_005274434.3 |
XP_005274491.1
|
9, 10 |
1311 |
|
XM_011546185.2 |
XP_011544487.1
|
6, 7 |
953 |
|
XM_017029919.1 |
XP_016885408.1
|
7, 8 |
1001 |
|
GenBank mRNA |
AK001090.1 |
- |
8, 9 |
1101 |
|
AK090498.1 |
- |
7, 8 |
1109 |
|
AK225770.1 |
- |
8, 9 |
1127 |
|
AK297805.1 |
- |
7, 8 |
1038 |
|
AK299231.1 |
- |
8, 9 |
1123 |
|
AK301844.1 |
- |
8, 9 |
1114 |
|
BC002508.2 |
- |
8, 9 |
1091 |
3051194 |
BC010089.2 |
- |
8, 9 |
1089 |
2988425 |
View Full Product Details