Gene Transcripts

Gene Symbol
CYBB
Entrez Gene ID
1536
Gene Name
cytochrome b-245 beta chain
Gene Aliases
AMCBX2, CGD, GP91-1, GP91-PHOX, GP91PHOX, IMD34, NOX2, p91-PHOX
Chromosome Location
Chr. X: 37780017 - 37813461 on Build GRCh38
UniGene ID
Hs.292356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000397.3 NP_000388.2 13 2650
GenBank mRNA BC032720.1 - 13 2636
X04011.1 - 13 2604

View Full Product Details