Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006893.2 | NP_008824.2 | 7 | 1015 | |
XM_005277509.3 | XP_005277566.1 | 7 | 930 | ||
XM_006711196.3 | XP_006711259.1 | 7 | 886 | ||
XM_011509257.2 | XP_011507559.1 | 7 | 930 | ||
XM_011509258.2 | XP_011507560.1 | 7 | 930 | ||
XM_017000532.1 | XP_016856021.1 | 7 | 886 | ||
GenBank mRNA | AB209364.1 | - | 1 | 1070 | |
AF159586.1 | - | 2 | 107 | ||
AF220417.2 | - | 7 | 923 | ||
AK001852.1 | - | 7 | 928 | ||
AK225259.1 | - | 7 | 909 | ||
AL162001.1 | - | 2 | 221 | ||
BC001585.1 | - | 7 | 913 | 3463026 | |
BC039134.1 | - | 7 | 866 | 4339127 | |
BC058905.1 | - | 7 | 880 | 6067337 |