Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000061.2 | NP_000052.1 | 11 | 1108 | |
NM_001287344.1 | NP_001274273.1 | 11 | 1264 | ||
NM_001287345.1 | NP_001274274.1 | 12 | 1273 | ||
GenBank mRNA | AK057105.1 | - | 12 | 1227 | |
AK225577.1 | - | 11 | 1070 | ||
AK309851.1 | - | 11 | 1250 | ||
AK314382.1 | - | 11 | 1075 | ||
BC109079.1 | - | 11 | 1040 | 40034287 | |
BC109080.1 | - | 11 | 1040 | 40034289 | |
KF241986.1 | - | 10 | 915 | ||
X58957.1 | - | 11 | 1047 |