Gene Transcripts
- Gene Symbol
- AP1G1
- Entrez Gene ID
- 164
- Gene Name
- adaptor related protein complex 1 gamma 1 subunit
- Gene Aliases
- ADTG, CLAPG1
- Chromosome Location
- Chr. 16: 71729000 - 71809073 on Build GRCh38
- UniGene ID
- Hs.461253
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001030007.1 |
NP_001025178.1
|
19, 20 |
2203 |
|
NM_001128.5 |
NP_001119.3
|
18, 19 |
2194 |
|
GenBank mRNA |
AB015317.1 |
- |
18, 19 |
1899 |
|
AK055122.1 |
- |
2, 3 |
814 |
|
AK128078.1 |
- |
21, 22 |
2749 |
|
AK225906.1 |
- |
19, 20 |
1962 |
|
AK314484.1 |
- |
18, 19 |
2199 |
|
AL110198.1 |
- |
2, 3 |
268 |
|
BC036283.1 |
- |
19, 20 |
2168 |
4158263 |
Y12226.1 |
- |
19, 20 |
1917 |
|
View Full Product Details