Gene Transcripts

Gene Symbol
GOLGB1
Entrez Gene ID
2804
Gene Name
golgin B1
Gene Aliases
GCP, GCP372, GOLIM1
Chromosome Location
Chr. 3: 121663199 - 121749908 on Build GRCh38
UniGene ID
Hs.213389
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256486.1 NP_001243415.1 13 4988
NM_001256487.1 NP_001243416.1 13 5016
NM_001256488.1 NP_001243417.1 12 4893
NM_004487.4 NP_004478.3 13 4973
XM_005247371.4 XP_005247428.1 13 5448
XM_005247372.4 XP_005247429.1 13 5024
XM_005247373.1 XP_005247430.1 12 4840
XM_006713587.1 XP_006713650.1 13 4963
XM_006713588.1 XP_006713651.1 13 4948
XM_006713589.1 XP_006713652.1 13 4945
XM_006713590.1 XP_006713653.1 12 4840
XM_006713591.1 XP_006713654.1 11 4723
XM_011512699.2 XP_011511001.1 13 4915
XM_017006189.1 XP_016861678.1 13 5433
XM_017006190.1 XP_016861679.1 13 5600
XM_017006191.1 XP_016861680.1 12 5325
XM_017006192.1 XP_016861681.1 13 5009
XM_017006193.1 XP_016861682.1 13 5006
XM_017006194.1 XP_016861683.1 12 4901
XM_017006195.1 XP_016861684.1 7 4870
GenBank mRNA AB371588.1 - 13 4961
AB593126.1 - 12 4867
BC065301.1 - 12 4764
D25542.1 - 13 4971
X75304.1 - 13 4961

View Full Product Details