Gene Transcripts

Gene Symbol
ZC3H6
Entrez Gene ID
376940
Gene Name
zinc finger CCCH-type containing 6
Gene Aliases
ZC3HDC6
Chromosome Location
Chr. 2: 112275594 - 112340063 on Build GRCh38
UniGene ID
Hs.190477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198581.2 NP_940983.2 10 1806
XM_006712519.2 XP_006712582.1 10 1813
GenBank mRNA AB111887.1 - 9 1252
AK127776.1 - 7 875
AK131416.1 - 10 1792
BC155540.1 - 9 1252
BC167154.1 - 9 1252

View Full Product Details