Gene Transcripts

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
Chromosome Location
Chr. 1: 200738905 - 200860707 on Build GRCh38
UniGene ID
Hs.23585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297708.1 NP_001284637.1 4, 5 912
NM_203459.2 NP_982284.1 4, 5 912
GenBank mRNA AK095640.1 - 4, 5 912
BC125230.1 - 4, 5 691 40124604

View Full Product Details