Gene Transcripts

Gene Symbol
ANKRD13D
Entrez Gene ID
338692
Gene Name
ankyrin repeat domain 13D
Gene Aliases
Chromosome Location
Chr. 11: 67289291 - 67302484 on Build GRCh38
UniGene ID
Hs.438673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207354.2 NP_997237.2 14 1752
NR_030767.1 15 1835
XM_011544985.1 XP_011543287.1 15 1842
XM_011544986.1 XP_011543288.1 16 1982
XM_011544987.2 XP_011543289.1 15 1816
XM_011544988.2 XP_011543290.1 10 1172
GenBank mRNA AK027313.1 - 7 1693
AK057681.1 - 7 2366
AK091614.1 - 2 2785
AK096277.1 - 14 1665
AK124721.1 - 15 1832
AK126438.1 - 13 1749
BC044239.1 - 4 723
BC110419.1 - 14 1725 5210878
BC121024.1 - 14 1626 40117420
BC121025.2 - 14 1626 40117423

View Full Product Details