Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159547.1 | NP_001153019.1 | 2, 3 | 1070 | |
NM_207406.3 | NP_997289.2 | 2, 3 | 1070 | ||
XM_017008185.1 | XP_016863674.1 | 1, 2 | 727 | ||
GenBank mRNA | AB126828.1 | - | 2, 3 | 1070 | |
AB126829.1 | - | 2, 3 | 1070 | ||
AK125953.1 | - | 2, 3 | 256 | ||
BC128592.1 | - | 2, 3 | 155 | 40127911 | |
BC128593.1 | - | 2, 3 | 155 | 40127915 |