Gene Transcripts
- Gene Symbol
- C11orf88
- Entrez Gene ID
- 399949
- Gene Name
- chromosome 11 open reading frame 88
- Gene Aliases
- Chromosome Location
- Chr. 11: 111514785 - 111537031 on Build GRCh38
- UniGene ID
- Hs.411600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128145.1 |
- |
6 |
1237 |
|
View Full Product Details