Gene Transcripts

Gene Symbol
Ccl11
Entrez Gene ID
20292
Gene Name
chemokine (C-C motif) ligand 11
Gene Aliases
Scya11, eotaxin
Chromosome Location
Chr. 11: 82057832 - 82062955 on Build GRCm38
UniGene ID
Mm.4686
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011330.3 NP_035460.1 2 301
GenBank mRNA AF128205.1 - 2 187
AF128206.1 - 2 187
AF128207.1 - 2 187
AF128208.1 - 2 187
AF128209.1 - 2 187
AK009307.1 - 2 284
AK010146.1 - 2 212
AW986913.1 - 2 301
BC027521.1 - 2 201 1527856
U26426.1 - 2 200
U40672.1 - 2 213

View Full Product Details