Gene Transcripts
- Gene Symbol
- Cdk17
- Entrez Gene ID
- 237459
- Gene Name
- cyclin-dependent kinase 17
- Gene Aliases
- 6430598J10Rik, Pctk2
- Chromosome Location
- Chr. 10: 93160876 - 93250688 on Build GRCm38
- UniGene ID
- Mm.217404
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC031778.1 |
- |
|
2038 |
|
BC049904.1 |
- |
|
2038 |
4501115 |
View Full Product Details