Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256005.1 | NP_001242934.1 | 11 | 1853 | |
NM_008620.3 | NP_032646.2 | 11 | 1790 | ||
GenBank mRNA | AK220567.1 | - | 11 | 1771 | |
BC141364.1 | - | 11 | 1741 | 9055977 | |
BC147434.1 | - | 11 | 1741 | 9055976 | |
EF494423.1 | - | 11 | 1853 | ||
EF494424.1 | - | 11 | 1824 |