Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010893.3 | NP_035023.3 | 3 | 576 | |
GenBank mRNA | AK077444.1 | - | 3 | 516 | |
AK145680.1 | - | 3 | 470 | ||
BC004666.1 | - | 3 | 450 | 3498197 | |
U93702.1 | - | 3 | 452 | ||
Y11412.1 | - | 3 | 455 |