Gene Transcripts

Gene Symbol
Ms4a2
Entrez Gene ID
14126
Gene Name
membrane-spanning 4-domains, subfamily A, member 2
Gene Aliases
FcRB, Fce1b, Fcer1b, Fcrbeta, Ms4a1, fcERI
Chromosome Location
Chr. 19: 11615517 - 11624034 on Build GRCm38
UniGene ID
Mm.329735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013516.2 NP_038544.1 5 540
GenBank mRNA BC139323.1 - 5 464
BC139324.1 - 5 464 8862346
J05019.1 - 5 464

View Full Product Details