Gene Transcripts

Gene Symbol
Ap3s1
Entrez Gene ID
11777
Gene Name
adaptor-related protein complex 3, sigma 1 subunit
Gene Aliases
Chromosome Location
Chr. 18: 46741876 - 46790826 on Build GRCm38
UniGene ID
Mm.312536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF084575.1 - 2 171
AK137489.1 - 2 192
AK152100.1 - 2 203
BC012656.1 - 2 182 4187607
BC108327.1 - 2 190 30713247
BC131985.1 - 2 172 40130262
BC145234.1 - 2 172

View Full Product Details