Gene Transcripts

Gene Symbol
Aqp4
Entrez Gene ID
11829
Gene Name
aquaporin 4
Gene Aliases
WCH4
Chromosome Location
Chr. 18: 15389394 - 15410977 on Build GRCm38
UniGene ID
Mm.250786
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009700.2 NP_033830.2 2 393
GenBank mRNA AF469168.1 - 2 467
AF469169.1 - 2 649
AK039777.1 - 2 395
AK044074.1 - 2 332
AK045357.1 - 1 44
AY484964.1 - 2 399
AY484965.1 - 3 583
AY484966.1 - 3 529
AY728043.1 - 2 413
AY728044.1 - 3 597
AY728045.1 - 2 525
AY803942.1 - 3 504
AY803943.1 - 4 634
AY803944.1 - 5 818
BB628835.1 - 2 394
BC024526.1 - 2 339 4985265
CD803766.1 - 2 518
CT010362.1 - 1 340
U33012.1 - 1 355
U48397.1 - 1 475
U48398.1 - 1 346
U48399.1 - 3 457
U88623.1 - 2 394

View Full Product Details