Gene Transcripts

Gene Symbol
PRKAB2
Entrez Gene ID
5565
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 2
Gene Aliases
Chromosome Location
Chr. 1: 147155106 - 147172589 on Build GRCh38
UniGene ID
Hs.50732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005399.4 NP_005390.1 3 384
XM_011509729.1 XP_011508031.1 3 384
XM_017001697.1 XP_016857186.1 3 384
GenBank mRNA AJ224538.1 - 2 200
AK292820.1 - 3 265
AK310982.1 - 3 265
BC053610.1 - 3 339 6091476
DA698245.1 - 3 384
KJ534919.1 - 2 200
KJ535025.1 - 2 200

View Full Product Details